EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology
Bioelectrical activity of brain in Rana tigrina (Daudin) in response to phosalone poisoning.
PMID 9440249
Abstract
An acute dose of phosalone, intraperitoneally injected to Rana tigrina alters the bioelectrical activity of the brain. The frequency levels of delta, theta, alpha, and beta waves remained similar in both control and experimental groups, while the amplitude of the waves was significantly decreased by 40 to 60% in the latter group. The total work done (TWD) was also significantly reduced. The above disturbance in the brain electrical activity may have correlation to the inhibition of cation-linked ATPase, as well as the cholinesterase activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36194
Phosalone, PESTANAL®, analytical standard
C12H15ClNO4PS2
PS682
Phosalone, analytical standard
C12H15ClNO4PS2