EMAIL THIS PAGE TO A FRIEND
Journal of clinical pharmacy and therapeutics
The effect of paroxetine on thiothixene pharmacokinetics.
PMID 9447478
Abstract
In this study healthy volunteers received thiothixene with and without a 3-day pretreatment with paroxetine to determine if paroxetine decreased the clearance of thiothixene. Ten healthy medication-free volunteers (4 women and 6 men, mean age 38 +/- 12 years) were randomized to receive a single 20 mg oral dose of thiothixene on two separate occasions. On one occasion thiothixene was given concurrently, and following 3 days of pre-treatment with oral paroxetine (20 mg/day). On the other occasion thiothixene was given without paroxetine pre-treatment. The two study days were separated by a minimum period of 2 weeks. On both study days, after the administration of thiothixene, 10 ml blood samples were collected over the next 72 h. None of the pharmacokinetic parameters of thiothixene were significantly altered by a 3-day treatment with paroxetine. It is likely that the CYP2D6 isoenzyme is not responsible for a high proportion of thiothixene clearance, but one cannot exclude the possibility that a longer paroxetine pretreatment might have caused some inhibition of thiothixene clearance.