EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Distribution of cyclic imide-transforming activity in microorganisms.
PMID 9453155
Abstract
Cyclic imide-transforming activity was found to be widely distributed in bacteria, yeast and molds. This activity was not correlated with cyclic ureide-transforming activity in bacteria, but there was some correlation in yeast and molds. These two activities are probably catalyzed by different enzymes in bacteria. Besides the well-known cyclic ureide transformation, cyclic imide transformation by microorganisms was common.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
533653
5-Methylhydantoin, 97%
C4H6N2O2