EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Assay of N8-acetylspermidine deacetylase.
PMID 9463820
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3658
N8-Acetylspermidine dihydrochloride, analytical standard
C9H21N3O · 2HCl