EMAIL THIS PAGE TO A FRIEND
FEBS letters
NMR studies of the conformation of thiocellobiose bound to a beta-glucosidase from Streptomyces sp.
PMID 9468315
Abstract
The conformation of 4-thiocellobiose bound to beta-glucosidase from Streptomyces sp. has been studied by 1H-NMR transferred nuclear Overhauser effect spectroscopy (TR-NOE). Thiocellobiose behaves as an inhibitor of this glucosidase when cellobiose is used as substrate. NOE measurements and molecular mechanics calculations have also been performed to estimate the probability distribution of conformers of thiocellobiose when free in solution. Experimental data show that, in contrast with the natural O-analogue, thiocellobiose presents three conformational families in the free state, namely syn, anti-psi and anti-phi, whilst only one of them (syn) is recognized by the enzyme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19608
Thiocellobiose, ≥98.0% (TLC)
C12H22O10S