EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Mechanistic studies on hydrotropic solubilization of nifedipine in nicotinamide solution.
PMID 9468644
Abstract
Nicotinamide is a hydrotropic agent that has been demonstrated to solubilize a wide variety of drugs through complexation. Past investigations on the potential interaction of nicotinamide with a solubilized drug have inadequately focused on aliphatic hydrotropes. This study examined the mechanism for the hydrotropic solubilization of nifedipine, a poorly water-soluble drug, in the aqueous solution of nicotinamide using not only nicotinamide analogues but also urea analogues as aliphatic hydrotropes. The values of stability constants, K1:2, at different temperatures in nicotinamide solution were determined by the phase solubility technique, and were utilized to estimate the thermodynamic parameters of complex formation between nifedipine and nicotinamide. The enthalpy change values suggested the participation of intermolecular forces other than hydrogen bonding in complexion. The aqueous solubility of nifedipine was measured in the presence of nicotinamide, urea and their analogues: N-methylnicotinamide, N,N-diethylnicotinamide, nipecotamide, methylurea, ethylurea and butylurea. The drug solubility increased in proportion to the amount of alkyl substituent on the amide nitrogen, and the solubilizing effect of butylurea was as high as that of nicotinamide. Furthermore, the relationship between the logarithmic drug solubilities in 1.0 M aqueous solutions of nicotinamide or urea analogues versus the logarithmic octanol-water partition coefficient values of ligands as an indication of hydrophobicity was found to be linear. The significant contributor to the hydrotropic solubilization of nifedipine with nicotinamide was therefore the ligand hydrophobicity rather than the aromaticity of the pyridine ring.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D98807
N,N-Diethylnicotinamide, 99%
C10H14N2O
B103004
N-Butylurea, 99%
C5H12N2O