EMAIL THIS PAGE TO A FRIEND
Chirality
Enantioseparation of atropisomeric 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate in capillary electrophoresis by using di- and oligosaccharides as chiral selectors: di- and oligosaccharide chiral selectors in capillary electrophoresis.
PMID 9470211
Abstract
Twelve different disaccharides and a series of noncyclic malto- and cello-oligosaccharides were used as chiral selectors in capillary electrophoresis (CE). Most saccharides resolved the enantiomers of atropisomeric 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate (BDHP) depending on the type (alpha or beta) and position of the linkage between monosaccharides. The effect of chain length of malto- and cello-oligosaccharides on enantioseparation of BDHP was also investigated. The nature of cations in background electrolytes affected significantly the separation of BDHP enantiomers.