EMAIL THIS PAGE TO A FRIEND
European journal of cancer (Oxford, England : 1990)
Combined effects of cisplatin and N,N-diethyl-2-[4-(phenylmethyl)phenoxy] ethanamine HCl on the growth of human ovarian cancer xenografts in nude mice.
PMID 9470857
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2571
DPPE hydrochloride, ≥98% (HPLC)
C19H25NO·HCl