EMAIL THIS PAGE TO A FRIEND
Soudni lekarstvi
[A sensitive method for detection of sympathomimetic amines in pentafluorobenzamide form in biological samples using gas chromatography].
PMID 9471730
Abstract
A toxicological method of sensitive and specific confirmation of methamphetamine and other primary and secondary amines in biological samples after extractive perfluorobenzoylation is described. The method is based on the principle of gas chromatography with parallel specific nitrogen detection and electron capture detection. The other alternative is the gas chromatography combined with mass spectrometry in chemical ionization mode. The method described allows the detection of methamphetamine and amphetamine in urine in concentrations below 10 ng/ml.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103772
2,3,4,5,6-Pentafluorobenzoyl chloride, 99%
C7ClF5O