EMAIL THIS PAGE TO A FRIEND
The Annals of pharmacotherapy
Use and abuse of flunitrazepam.
PMID 9475831
Abstract
Flunitrazepam is not available in the US, and it is presently illegal to import it. Flunitrazepam has no therapeutic advantage over benzodiazepines presently marketed in the US. Its quick onset of sedation, amnesic properties, and additive effects with alcohol have led to its reputation as a party drug, club drug, or date rape drug. Several measures have been undertaken to curb its illicit use. It is unclear whether flunitrazepam poses a greater public health risk than other benzodiazepines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0190000
Flunitrazepam, European Pharmacopoeia (EP) Reference Standard
C16H12FN3O3