EMAIL THIS PAGE TO A FRIEND
Przeglad lekarski
Factors influencing the efficacy of obidoxime in organophosphate pesticides poisoning.
PMID 9478082
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34234
Demeton-S-methyl solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C6H15O3PS2