EMAIL THIS PAGE TO A FRIEND
The Biochemical Journal
3-Nitrotyrosine in the proteins of human plasma determined by an ELISA method
PMID 9480893
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-284
Anti-Nitrotyrosine Antibody, clone, Upstate®, from rabbit