EMAIL THIS PAGE TO A FRIEND
Northwestern dental research
An in vitro model for studying oral bone loss.
PMID 9487923
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3671 Parathyroid Hormone Fragment 1-34 bovine, ≥97% (HPLC), powder
C183H288N54O50S2