EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
Modulation of human glutamate transporter activity by phorbol ester.
PMID 9489718
Abstract
Termination of synaptic glutamate transmission depends on rapid removal of glutamate by neuronal and glial high-affinity transporters. Molecular biological and pharmacological studies have demonstrated that at least five subtypes of Na+-dependent mammalian glutamate transporters exist. Our study demonstrates that Y-79 human retinoblastoma cells express a single Na+-dependent glutamate uptake system with a Km of 1.7 +/- 0.42 microM that is inhibited by dihydrokainate and DL-threo-beta-hydroxyaspartate (IC50 = 0.29 +/- 0.17 microM and 2.0 +/- 0.43 microM, respectively). The protein kinase C activator phorbol 12-myristate 13-acetate caused a concentration-dependent inhibition of glutamate uptake (IC50 = 0.56 +/- 0.05 nM), but did not affect Na+-dependent glycine uptake significantly. This inhibition of glutamate uptake resulted from a fivefold decrease in the transporter's affinity for glutamate, without significantly altering the Vmax. 4Alpha-phorbol 12,13-didecanoate, a phorbol ester that does not activate protein kinase C, did not alter glutamate uptake significantly. The phorbol 12-myristate 13-acetate-induced inhibition of glutamate uptake was reversed by preincubation with staurosporine. The biophysical and pharmacological profile of the human glutamate transporter expressed by the Y-79 cell line indicates that it belongs to the dihydrokainate-sensitive EAAT2/GLT-1 subtype. This conclusion was confirmed by western blot analysis. Protein kinase C modulation of glutamate transporter activity may represent a mechanism to modulate extracellular glutamate and shape postsynaptic responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148946
DL-2-Methylglutamic acid hemihydrate, 98%
C6H11NO4 · 0.5H2O