EMAIL THIS PAGE TO A FRIEND
The Journal of physiology
alpha2-Adrenoceptor-mediated potassium currents in acutely dissociated rat locus coeruleus neurones.
PMID 9490817
Abstract
1. The noradrenaline (NA)-activated response was investigated in neurones acutely dissociated from the rat locus coeruleus (LC) using nystatin-perforated, conventional whole-cell and inside-out patch recording modes under current- and voltage-clamp conditions. 2. Under current-clamp conditions, NA hyperpolarized the LC neurones, abolishing the spontaneous action potentials. In voltage-clamp studies, NA induced an inwardly rectifying K+ current (INA) in a concentration-dependent manner with a half-maximum effective concentration of 2.2 x 10(-7) M. 3. INA was mimicked by the alpha2-agonist UK14304 but was inhibited by either the alpha2B/alpha2C antagonist ARC239 or the alpha1- and alpha2B/alpha2C antagonist prazosin, suggesting the contribution of alpha2B/alpha2C adrenoceptors. 4. INA was inhibited by the intracellular application of GDPbetaS but fully activated by intracellular perfusion of GTPgammaS. 5. In the inside-out recording mode, the application of GTP to the cytoplasmic side of the patch membrane markedly enhanced the open probability of the NA-activated single channels which represented the inwardly rectifying properties. 6. These results indicate that the activation of alpha2B/alpha2C adrenoceptors coupled with GTP-binding protein directly activates the inwardly rectifying K+ currents in rat LC neurones, thus resulting in a decrease in the spontaneous firing activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5736
ARC 239 dihydrochloride hydrate, ≥98% (HPLC)
C24H29N3O3·2HCl · xH2O