EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Localization and functional analysis of the substrate specificity/catalytic domains of human M-form and P-form phenol sulfotransferases.
PMID 9497349
Abstract
Human monoamine (M)-form and simple phenol (P)-form phenol sulfotransferases (PSTs), which are greater than 93% identical in their primary sequences, were used as models for investigating the structural determinants responsible for their distinct substrate specificity and other enzymatic properties. A series of chimeric PSTs were constructed by reciprocal exchanges of DNA segments between cDNAs encoding M-form and P-form PSTs. Functional characterization of the recombinant wild-type M-form, P-form, and chimeric PSTs expressed in Escherichia coli and purified to homogeneity revealed that internal domain-spanning amino acid residues 84-148 contain the structural determinants for the substrate specificity of either M-form or P-form PST. Data on the kinetic constants (Km, Vmax, and Vmax/Km) further showed the differential roles of the two highly variable regions (Region I spanning amino acid residues 84-89 and Region II spanning amino acid residues 143-148) in substrate binding, catalysis, and sensitivity to the inhibition by 2,6-dichloro-4-nitrophenol. In contrast to the differential sulfotransferase activities of M-form and P-form PSTs toward dopamine and p-nitrophenol, the Dopa/tyrosine sulfotransferase activities were found to be restricted to M-form, but not P-form, PST. Furthermore, the variable Region II of M-form PST appeared to play a predominant role in determining the Dopa/tyrosine sulfotransferase activities of chimeric PSTs. Kinetic studies indicated the role of manganese ions in dramatically enhancing the binding of D-p-tyrosine to wild-type M-form PST. Taken together, these results pinpoint unequivocally the sequence encompassing amino acid residues 84-148 to be the substrate specificity/catalytic domain of both M-form and P-form PSTs and indicate the importance of the variable Regions I and II in determining their distinct enzymatic properties.