EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Melanin biosynthesis in Cryptococcus neoformans.
PMID 9515929
Abstract
Pigment production by Cryptococcus neoformans is virulence associated. Dopamine- and 3,4-dihydroxyphenylalanine-melanin products were identified after acidic permanganate oxidation, alkaline hydrogen peroxide oxidation, or hydrolysis with hydriodic acid. These data provide direct chemical evidence for the formation of eumelanin polymers by catecholamine oxidation by laccase alone followed by oxidative coupling of dihydroxyindole.