EMAIL THIS PAGE TO A FRIEND
Diabetes
4-Hydroxyisoleucine: a novel amino acid potentiator of insulin secretion.
PMID 9519714
Abstract
We report the characterization of a new insulinotropic compound, 4-hydroxyisoleucine. This amino acid has been extracted and purified from fenugreek seeds, which are known in traditional medicine for their antidiabetic properties. 4-Hydroxyisoleucine increases glucose-induced insulin release, in the concentration range of 100 micromol/l to 1 mmol/l, through a direct effect on isolated islets of Langerhans from both rats and humans. The stimulating effect of 4-hydroxyisoleucine was strictly glucose dependent; indeed, ineffective at low (3 mmol/l) or basal (5 mmol/l) glucose concentrations, the amino acid potentiated the insulin secretion induced by supranormal (6.6-16.7 mmol/l) concentrations of glucose. In addition, in the isolated perfused rat pancreas, we could show 1) that the pattern of insulin secretion induced by 4-hydroxyisoleucine was biphasic, 2) that this effect occurred in the absence of any change in pancreatic alpha- and delta-cell activity, and 3) that the more glucose concentration was increased, the more insulin response was amplified. Moreover, 4-hydroxyisoleucine did not interact with other agonists of insulin secretion (leucine, arginine, tolbutamide, glyceraldehyde). Therefore, we conclude that 4-hydroxyisoleucine insulinotropic activity might, at least in part, account for fenugreek seeds' antidiabetic properties. This secretagogue may be considered as a novel drug with potential interest for the treatment of NIDDM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50118
(4S)-4-Hydroxy-L-isoleucine, from fenugreek seed, ≥98.0% (TLC)
C6H13NO3