EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Protein kinase C disrupts cannabinoid actions by phosphorylation of the CB1 cannabinoid receptor.
PMID 9526000
Abstract
We have found that phosphorylation of a G-protein-coupled receptor by protein kinase C (PKC) disrupts modulation of ion channels by the receptor. In AtT-20 cells transfected with rat cannabinoid receptor (CB1), the activation of an inwardly rectifying potassium current (Kir current) and depression of P/Q-type calcium channels by cannabinoids were prevented by stimulation of protein kinase C by 100 nM phorbol 12-myristate 13-acetate (PMA). In contrast, activation of Kir current by somatostatin was unaffected, and inhibition of calcium channels was only modestly attenuated. The possibility that PKC acted by phosphorylating CB1 receptors was confirmed by demonstrating that PKC phosphorylated a single serine (S317) of a fusion protein incorporating the third intracellular loop of CB1. Mutating this serine to alanine did not affect the ability of CB1 to modulate currents, but it eliminated disruption by PMA, demonstrating that PKC can disrupt ion channel modulation by receptor phosphorylation.