EMAIL THIS PAGE TO A FRIEND
Revista de neurologia
[Tardive dystonia following the administration of clebopride].
PMID 9528065
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8289
Clebopride maleate salt, ~95%
C20H24ClN3O2 · C4H4O4