EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Determination of the main tropane alkaloids from transformed Hyoscyamus muticus plants by capillary zone electrophoresis.
PMID 9535182
Abstract
A capillary zone electrophoretic method (CZE) was developed using an uncoated fused silica capillary for the separation and determination of the main tropane alkaloids. The applicability of the developed method for analysis of plant samples was examined by analyzing samples of transgenic Egyptian henbane Hyoscyamus muticus (L.) plants. A simple 40 mM phosphate buffer at pH 7.8 using a voltage of 20 kV was found the best for this purpose. The main tropane alkaloids, atropine and scopolamine as well as nor-(-)-scopolamine, and tropic acid, the precursor of tropane alkaloids, could be separated in less than 13 min. The linear concentration range for atropine was 5.00-140 microg ml(-1), for scopolamine 7.50-210 microg ml(-1) and for tropic acid 2.50-70.0 microg ml(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T89206
Tropic acid, 98%
C9H10O3