EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Functional analysis of the Huntington's disease (HD) gene promoter.
PMID 9536082
Abstract
The basis for the highly specific neuronal vulnerability seen in Huntington's disease (HD) has not been determined. Recent studies have demonstrated that variation in HD protein expression occurs in the striatum, with affected regions showing increased HD immunoreactivity. Experiments in HD and SCA1 transgenic mice suggest a correlation between phenotypic severity and expression of the mutant transgene. To gain insights into control of HD gene expression, and to investigate the possibility of cell-cell differences in transcription, we have analysed the 5' upstream region of the HD gene in a neuronal (SK-N-SH) and a non-neuronal (JEG3) cell line. Reporter gene assays demonstrated the presence of a key positive-acting region apparently arising from two Sp1 sites in a tandem repeat acting synergistically. This site is polymorphic, and a single Sp1 site is associated with reduced levels of transcription. These experiments also reveal differences in control of expression between neuronal and non-neuronal cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
JEG3, 92120308