EMAIL THIS PAGE TO A FRIEND
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova
[Peptide modulation of vagal sinus arrhythmia in cats].
PMID 9541964
Abstract
In anesthetised cats, following vagal stimulation with short bursts of pulses, a narrow zone was identified within the cardiac cycle where even a small change of the vagal burst position produced sharp alterations in the magnitude of the chronotropic effect. Somatostatin diminished the size of this zone whereas its antagonist exerted an opposite effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4801
Cyclo(7-aminoheptanoyl-Phe-D-Trp-Lys-Thr[Bzl]), ≥95% (HPLC), powder
C44H57N7O6