EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Effects of oxidants on membrane potential, K+ and Ca2+ currents of mouse pancreatic B-cells.
PMID 9544294
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158194
2,2′-Dithiobis(5-nitropyridine), 96%
C10H6N4O4S2