EMAIL THIS PAGE TO A FRIEND
Acta chemica Scandinavica (Copenhagen, Denmark : 1989)
Synthesis of meiosis-activating sterols containing fluorine.
PMID 9550087
Abstract
It is documented that specific types of sterol play a major role in the resumption of meiosis in oocytes from mice in vitro. 4,4-Dimethyl-5 alpha-cholesta-8,14,24-trien-3 beta-ol (FF-MAS) isolated from human follicular fluid and 4,4-dimethyl-5 alpha-cholesta-8,24-dien-3 beta-ol (T-MAS) isolated from bull testicular tissue, have been shown to activate (promote) meiosis in vitro. In order to evaluate the biological activity and stability of such compounds, new demethylsterol derivatives have been synthesised. Using diethylaminosulfur trifluoride (DAST) it was possible to synthesise selected delta 8, delta 14 sterols with mono and difluoro substitution at C3.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235253
(Diethylamino)sulfur trifluoride, 95%
C4H10F3NS