EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Availability of O2 as a substrate in the cytoplasm of bacteria under aerobic and microaerobic conditions.
PMID 9555896
Abstract
The growth rates of Pseudomonas putida KT2442 and mt-2 on benzoate, 4-hydroxybenzoate, or 4-methylbenzoate showed an exponential decrease with decreasing oxygen tensions (partial O2 tension [pO2] values). The oxygen tensions resulting in half-maximal growth rates were in the range of 7 to 8 mbar of O2 (corresponding to 7 to 8 microM O2) (1 bar = 10(5) Pa) for aromatic compounds, compared to 1 to 2 mbar for nonaromatic compounds like glucose or succinate. The decrease in the growth rates coincided with excretion of catechol or protocatechuate, suggesting that the activity of the corresponding oxygenases became limiting. The experiments directly establish that under aerobic and microaerobic conditions (about 10 mbar of O2), the diffusion of O2 into the cytoplasm occurs at high rates sufficient for catabolic processes. This is in agreement with calculated O2 diffusion rates. Below 10 mbar of O2, oxygen became limiting for the oxygenases, probably due to their high Km values, but the diffusion of O2 into the cytoplasm presumably should be sufficiently rapid to maintain ambient oxygen concentrations at oxygen tensions as low as 1 mbar of O2. The consequences of this finding for the availability of O2 as a substrate or as a regulatory signal in the cytoplasm of bacterial cells are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T36803
p-Toluic acid, 98%
C8H8O2