EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
The roles of the basic residues in the prosegment of aspergillopepsinogen I.
PMID 9561225
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2143 Protease from Aspergillus saitoi, Type XIII, ≥0.6 unit/mg solid