EMAIL THIS PAGE TO A FRIEND
Fortschritte der Medizin
[Contraception per computer. Hormone system persona--results of studies in Germany].
PMID 9577075
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2127
β-Estradiol 3-(β-D-glucuronide) sodium salt
C24H31NaO8