EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Carbachol-induced hydrolysis of phospholipids in hippocampal slices may be mediated in part by subsequent activation of metabotropic glutamate receptors.
PMID 9580390
Abstract
We observed that AP-3, an antagonist of metabotropic glutamate receptors, reduced carbachol-induced hydrolysis of phospholipids in hippocampal slices. This inhibition could be explained in different ways, e.g.: 1) AP-3 acts also as antagonist of muscarinic receptors mediating the hydrolysis of phospholipids induced by carbachol, 2) Carbachol induces the release of glutamate which, by activating metabotropic glutamate receptors, leads to additional hydrolysis of phospholipids. The aim of this work was to test these possibilities. It is shown that AP-3 reduces carbachol-induced hydrolysis of phospholipids in hippocampal slices but not in cerebellar neurons at 10-14 days of culture, when these cells are not able to induce hydrolysis of phospholipids following activation of metabotropic glutamate receptors. It is also shown that carbachol induces a release of [3H]aspartate in hippocampal slices. The results reported suggest that the hydrolysis of phospholipids induced by carbachol in hippocampal slices would have two components. One part would be due to direct activation by carbachol of muscarinic receptors associated to activation of phospholipase C. This part would not be inhibited by AP-3. The second part would be due to subsequent release of glutamate and activation of metabotropic glutamate receptors. This part would be inhibited by AP-3.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4910
DL-2-Amino-3-phosphonopropionic acid
C3H8NO5P