EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Eastern Cooperative Oncology Group randomized trials of observation versus maintenance therapy for patients with metastatic breast cancer in complete remission following induction treatment.
PMID 9586877
Abstract
To investigate the value of maintenance treatment for patients with metastatic breast cancer whose disease is in complete remission (CR). One hundred ninety-five women (141 eligible) whose disease was in CR or in CR except for bone metastases following six cycles (6 months) of doxorubicin-containing induction treatment were randomized to receive cyclophosphamide, methotrexate, fluorouracil, prednisone, tamoxifen, and halotestin [CMF(P)TH] or observation. In a previous pilot study, patients in CR after 24 months of induction treatment were randomized to continue chemotherapy for 4 more years or stop chemotherapy. Among patients randomized to CMF(P)TH, life-threatening toxicity included leukopenia in 3%, thrombocytopenia in 3%, cardiac in 2%, and diabetes in 1%. The median time to relapse from randomization was 18.7 months on CMF(P)TH and only 7.8 months on observation (P < .0001). The median time to death was 32.2 months on CMF(P)TH and 28.7 months on observation (P=.74). Similar results were seen in the pilot study (median time to relapse, 12.6 and 6.4 months; median survival, 37.7 and 24.2 months; study too small for statistical significance). Maintenance treatment was always the most significant covariate in time-to-relapse models. There is definite toxicity associated with CMF(P)TH maintenance treatment. When CR was obtained on induction, maintenance treatment with CMF(P)TH was never significant in survival models. However, maintenance treatment was always the most significant covariate in the time-to-relapse models, which motivates its consideration for appropriately informed patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7751
Fluoxymesterone, solid (photosensitive)
C20H29FO3