EMAIL THIS PAGE TO A FRIEND
Neurosurgery
The in vivo metabolic pattern of low-grade brain gliomas: a positron emission tomographic study using f-18-fluorodeoxyglucose and c-11-L-methylmethionine.
PMID 9588575