EMAIL THIS PAGE TO A FRIEND
Ryoikibetsu shokogun shirizu
[Sarcosine dehydrogenase deficiency].
PMID 9590030
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7672
Sarcosine, BioXtra
C3H7NO2
131776
Sarcosine, 98%
C3H7NO2
84532
Sarcosine, crystallized, ≥98.0% (T)
C3H7NO2