EMAIL THIS PAGE TO A FRIEND
Methods and findings in experimental and clinical pharmacology
Paclitaxel, bropirimine and linomide: effect on growth inhibition in a murine prostate cancer model by different growth regulatory mechanisms.
PMID 9604852
Abstract
Paclitaxel, bropirimine and linomide therapy was evaluated in a murine prostate cancer model. All drugs were effective in impeding tumor growth but the mechanisms of action varied. Paclitaxel inhibited bcl-2 expression suggesting an apoptotic mechanism. Bropirimine, while inhibiting bcl-2 expression also significantly depressed tumor necrosis factor-alpha (TNF-alpha) expression. In the bropirimine treated group there was also a correlation between angiogenesis and cyclin D expression. Finally, linomide significantly decreased angiogenesis. Since the mechanism of action of these drugs differ, combining them at lower doses might maintain therapeutic efficacy while reducing toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0218
Bropirimine, ≥98% (HPLC)
C10H8BrN3O