EMAIL THIS PAGE TO A FRIEND
Pharmacy world & science : PWS
Torsade de pointes after pipamperone intoxication.
PMID 9618739
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3246
Pipamperone dihydrochloride, ~99% (HPLC), powder
C21H30N3O2F · 2HCl