EMAIL THIS PAGE TO A FRIEND
The Yale journal of biology and medicine
The use of synthetic analogues of Arg-Gly-Asp (RGD) and soluble receptor of tumor necrosis factor to prevent acute and chronic experimental liver injury.
PMID 9626759
Abstract
In chronic viral hepatitis, autoimmune hepatitis, and some chronic cholestatic liver diseases, T-lymphocytes serve as effector cells of the immunostimulatory processes. Cellular interactions of immune cells with extracellular matrix (ECM) components are regulated primarily via the beta 1 subfamily of integrin receptors. The target epitope of several such integrin receptors is the Arg-Gly-Asp (RGD) sequence, a cell adhesion motif shared by several matrix-associated adhesive glycoproteins. We review the use of synthetic nonpeptidic analogues of RGD and of soluble receptor of tumor necrosis factor (TNF)-alpha in the prevention of immune-mediated, concanavalin A-induced liver damage in mice and of RGD analogues in inhibiting the development of liver cirrhosis in rats. The concanavalin A-induced elevation of serum transaminases and TNF-alpha, and the infiltration of liver tissue by inflammatory cells, were inhibited by pretreatment of the mice with the synthetic RGD mimetics and soluble TNF receptor. In rats, the progression of thioacetamide-induced liver cirrhosis was markedly inhibited by the coadministration of the RGD mimetic SF-6,5. The compounds described here may be examined therapeutically for pathological conditions in the liver, manifested as necroinflammation, cholestasis and fibrosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8052
Arg-Gly-Asp, ≥97% (TLC)
C12H22N6O6