EMAIL THIS PAGE TO A FRIEND
Neurology
High prevalence of parkinsonism after occupational exposure to lead-sulfate batteries.
PMID 9633752
Abstract
Seven of nine postal workers exposed to lead-sulfate batteries over a period of up to 30 years developed parkinsonian symptoms. One of the remaining two showed left-hand bradykinesia and one was not available for examination. The high prevalence and cause of parkinsonism in these patients remains unexplained. Lead intoxication may play a role in the occurrence of parkinsonian symptoms, but involvement of sulfate and other sulfur compounds must also be considered.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
307734
Lead(II) sulfate, 98%
O4PbS
254258
Lead(II) sulfate, 99.995% trace metals basis
O4PbS