EMAIL THIS PAGE TO A FRIEND
Ryoikibetsu shokogun shirizu
[Methionine malabsorption syndrome].
PMID 9645139
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54917
(R)-2-Hydroxybutyric acid, ≥97.0% (T)
C4H8O3