EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
cDNA cloning, expression, and characterization of the human bifunctional ATP sulfurylase/adenosine 5'-phosphosulfate kinase enzyme.
PMID 9648242
Abstract
A cDNA encoding the human bifunctional ATP sulfurylase/adenosine 5'-phosphosulfate (APS) kinase was cloned and sequenced. The enzyme contains an APS kinase domain in its N-terminal portion and an ATP sulfurylase domain in its C-terminal portion. Recombinant full-length enzyme and its constituent APS kinase and ATP sulfurylase domains were individually expressed, purified, and shown to have their respective enzymatic activities.