EMAIL THIS PAGE TO A FRIEND
The Journal of allergy and clinical immunology
Occupational asthma caused by tetrachlorophthalic anhydride: a 12-year follow-up.
PMID 9648696
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131865
Tetrachlorophthalic anhydride, 96%
C8Cl4O3