EMAIL THIS PAGE TO A FRIEND
Mutation research
Isolation of a novel metabolizing system enriched in phase-II enzymes for short-term genotoxicity bioassays.
PMID 9651530
Abstract
Murine S9 liver fractions isolated from mice fed 7.5 g kg-1 2(3)-tert-Butyl-4-hydroxyanisole (BHA) for 3 weeks were tested to determine: (a) the profile of both phase-I and phase-II xenobiotic metabolizing enzymes; (b) their ability to induce in vitro covalent binding of some precarcinogens to calf thymus DNA; and (c) their activation in a standard genetic toxicology assay. With regard to phase-I pathway, the S9 fraction expressed various cytochrome P-450-(CYP) (classes 1A1, 1A2, 2B1, 2E1, and 3A)-dependent biotransformation enzymes at levels comparable with those present in murine control liver. For post-oxidative enzymes, the S9 expressed high levels of glutathione S-transferases (up to 12-fold increase), glutathione S-epoxide-transferase (up to 2.6-fold), UDP-glucuronosyl transferase (up to 5.3-fold) and epoxide hydrolase (up to 2.6-fold) activities, as compared to untreated mice. The in vitro DNA binding of the precarcinogenic agents [14C]-1,4-dichlorobenzene, [14C]-1,2-dichlorobenzene and [14C]-1,4-dibromobenzene, mediated by BHA-induced cytosol and/or microsomal preparation, showed an increase in specific activity comparable to that observed with phase-I (PB/beta NF) induced S9. In some instances, covalent binding was even more elevated using the BHA-induced systems as compared with traditional S9 fractions. For example, cytosol derived from BHA-administered mice was able to induce a significant binding to calf thymus DNA up to 26.2-fold increase for [14C]-1,4-dichlorobenzene, while cytosol from PB/beta NF was not. A high mutagenic response on diploid D7 strain of Saccharomyces cerevisiae as exemplified by a marked induction of mitotic gene conversion and point (reverse) mutation confirmed that BHA-derived S9 fractions activate precarcinogens to final genotoxins. Because a number of chemicals are activated by either oxidative or post-oxidative enzymes, the use of metabolizing biosystems, with an enhanced phase-II pathway, together with classical S9 fractions, can improve the sensitivity of the assay in detecting unknown genotoxins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D39029
1,4-Dibromobenzene, 98%
C6H4Br2