EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Oxindole in pathogenesis of hepatic encephalopathy.
PMID 9652676
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9808
2-Oxindole, 97%
C8H7NO