EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Insect growth regulators. Analogues of TH-6038 and TH-6040.
PMID 965594
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
544264
2,6-Difluorobenzenesulfonyl chloride, 97%
C6H3ClF2O2S