EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of alpha-glucosidase inhibitors: kojibiose-type pseudo-disaccharides and a related pseudotrisaccharide.
PMID 9658566
Abstract
Two kojibiose-type pseudo-disaccharides and a trisaccharide, containing a 5-amino-1,2,3,4-cyclopentanetetrol derivative or valienamine, linked by way of nitrogen bridges to the sugar residues, have been designed and synthesized as processing alpha-glucosidase I inhibitors. Synthesis of the pseudo-disaccharides was carried out starting from the coupling products of the sugar isothiocyanates and an aminocyclitol, respectively, by cyclization with mercury(II) oxide to the cyclic isoureas and subsequent deprotection. Pseudokojibiose was prepared in a poor yield by reaction of a protected valienamine and a sugar epoxide, followed by deprotection. Although the pseudooligosaccharides are all strong inhibitors of alpha-glucosidase (baker's yeast), they did not have any inhibitory potency against either sucrase isomaltase (rat intestine) or processing alpha-glucosidase (rat liver microsomes).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K4769
Kojibiose, ≥98% (HPLC)
C12H22O11