EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Targeting, capture, and stabilization of microtubules at early focal adhesions.
PMID 9660872
Abstract
By co-injecting fluorescent tubulin and vinculin into fish fibroblasts we have revealed a "cross talk" between microtubules and early sites of substrate contact. This mutuality was first indicated by the targeting of vinculin-rich foci by microtubules during their growth towards the cell periphery. In addition to passing directly over contact sites, the ends of single microtubules could be observed to target several contacts in succession or the same contact repetitively, with intermittent withdrawals. Targeting sometimes involved side-stepping, or the major re-routing of a microtubule, indicative of a guided, rather than a random process. The paths that microtubules followed into contacts were unrelated to the orientation of stress fiber assemblies and targeting occurred also in mouse fibroblasts that lacked a system of intermediate filaments. Further experiments with microtubule inhibitors showed that adhesion foci can: (a) capture microtubules and stabilize them against disassembly by nocodazole; and (b), act as preferred sites of microtubule polymerization, during either early recovery from nocodazole, or brief treatment with taxol. From these and other findings we speculate that microtubules are guided into substrate contact sites and through the motor-dependent delivery of signaling molecules serve to modulate their development. It is further proposed this modulation provides the route whereby microtubules exert their influence on cell shape and polarity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1274
Poly-L-lysine hydrobromide, mol wt 70,000-150,000 by viscosity