EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Screening of monoclonal antibodies using antigens labeled with acetylcholinesterase.
PMID 9664363
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9043
S-Acetylthioglycolic acid N-hydroxysuccinimide ester, ≥95% (TLC), powder
C8H9NO5S