EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Isobutyl nitrite liberates nitric oxide which is not responsible for the immunotoxicity of the inhalant.
PMID 9666279
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
327190
Isobutyl nitrite, 95%
C4H9NO2