EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Molecular cloning, expression, and characterization of human bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase and its functional domains.
PMID 9668121
Abstract
The universal sulfonate donor, 3'-phosphoadenosine 5'-phosphosulfate (PAPS), is synthesized by the concerted action of ATP sulfurylase and adenosine 5'-phosphosulfate (APS) kinase, which in animals are fused into a bifunctional protein. The cDNA for human PAPS synthase (hPAPSS) along with polymerase chain reaction products corresponding to several NH2- and COOH-terminal fragments were cloned and expressed in COS-1 cells. A 1-268-amino acid fragment expressed APS kinase activity, whereas a 220-623 fragment evinced ATP sulfurylase activity. The 1-268 fragment and full-length hPAPSS (1-623) exhibited hyperbolic responses against APS substrate with equivalent Km values (0.6 and 0.4 microM, respectively). The 1-268 fragment demonstrated Michaelis-Menten kinetics against ATP as substrate (Km 0.26 mM); however, full-length hPAPSS exhibited a sigmoidal response (apparent Km 1.5 mM) suggesting cooperative binding. Catalytic efficiency (Vmax/Km) of the 1-268 fragment was 64-fold higher than full-length hPAPSS for ATP. The kinetic data suggest that the COOH-terminal domain of hPAPSS exerts a regulatory role over APS kinase activity located in the NH2-terminal domain of this bifunctional protein. In addition, the 1-268 fragment and full-length hPAPSS were overexpressed in Escherichia coli and column purified. Purified full-length hPAPSS, in contrast to the COS-1 cell-expressed cDNA construct, exhibited a hyperbolic response curve against ATP suggesting that hPAPSS is perhaps modified in vivo.