EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Butyrate trials.
PMID 9668533
Abstract
The aims of this study were to ascertain tolerability, safety and efficacy of oral isobutyramide (150 mg/kg bw/day) in stimulating fetal hemoglobin production in twelve thalassemia intermedia patients. Patients were treated for 28 days and followed for a further 28 days. Efficacy was monitored by non-alpha/alpha globin chain ratio and percentage of HbF. Five patients experienced increases of non-alpha/alpha ratio ranging between 5.3 and 100% at the end of treatment. Five patients show an increase of HbF ranging between 4.4 and 26%. Their HbF% continues to increase during follow-up period. The analysis of variance for HbF showed a time effect close to significance both in treatment period (p = 0.06) and in follow-up period (p = 0.08). Moreover, to evaluate a possible erythropoietic modification, serum Erythropoietin (sEpo) and serum Transferrin Receptor (sTfR) were evaluated. Serum Epo and sTfR levels were significantly increased during treatment (p < 0.05 vs baseline).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
144436
Isobutyramide, 99%
C4H9NO