EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of some divalent O- and S-glycosides of galabiose and globotriose.
PMID 9675366
Abstract
Derivatives of galabiose (alpha-D-Galp-(1-->4)-D-Galp) and globotriose (a-D-Galp-(1-->4)-beta-D-Galp-(1-->4)-D-Glcp) were coupled to various 1,2- and 1,3-dihydroxymethyl- and dimercapto-methylbenzenes to give the corresponding divalent glycosides, potentially useful as inhibitors of bacterial adhesion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G9287
Globotriose
C18H32O16