EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Targeting Holliday junctions by the RecG branch migration protein of Escherichia coli.
PMID 9677403
Abstract
The RecG protein of Escherichia coli is a junction-specific DNA helicase that drives branch migration of Holliday intermediates in genetic recombination and DNA repair. The reaction was investigated using synthetic X-junctions. RecG dissociates X-junctions to flayed duplex products, although DNA unwinding of the heterologous arms is limited to